Learn More
Thermo Scientific™ Culti-Loops™ Alcaligenes faecalis subsp. faecalis ATCC® 8750™
Catalog No. R4601006 Shop All Thermo Scientific Products
Change view
Click to view available options
Alcaligenes faecalis subsp. faecalis; ATCC 8750
Catalog No.
R4601006 Alcaligenes faecalis subsp. faecalis; ATCC 8750
1 options
This item is not returnable. View return policy
Catalog No. R4601006 Supplier Thermo Scientific™ Supplier No. R4601006
Add to Cart
Add to Cart
This item is not returnable. View return policy
Culti-Loops™ are disposable bacteriological loops containing stabilised viable microorganisms.
Use Thermo Scientific™ Culti-Loops™ Alcaligenes faecalis subsp. faecalis ATCC™ 8750™ in the performance testing of culture media, stains, diagnostic kits, and reagents, for the maintenance of stock cultures and in the evaluation of bacteriological procedures. Culti-Loops™ are ready to use, disposable bacteriological loops containing stabilised viable microorganisms.
The film in each loop is made from a gelatine formulation and then dried by special processing. To rehydrate the film, the loops must come into contact with both warmth and moisture. Most organisms grow in 24–48 hours under the proper conditions. However, some exhibit a considerable lag phase and should be incubated for an additional 24 hours.
  • Ready-to-use – Prepared disposable bacteriological loops with viable microorganisms
  • Easy-to-store – Store at 2-8°C
There are two methods such as direct streak method and indirect streak method which may be used for inoculation. The appropriate method is used for the selected microorganism.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
TRUSTED_SUSTAINABILITY
Specifications
ATCC® 8750TM*
Description Alcaligenes faecalis subsp. faecalis
Product Type Culti-Loops Quality Control Organism
Target Organism Class Gram-negative Rods
Test Type Qualitative QC
Quantity 5 Loops/Pack
Strain Designations WDCM00103, 16 [NCIB 8156]
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.