Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Culti-Loops™ API™ 20E QC Set

Catalog No. r4651003
Change view
Click to view available options
Quantity:
5 Loops/Organism
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R4651003 5 Loops/Organism
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. R4651003 Supplier Thermo Scientific™ Supplier No. R4651003
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.

Thermo Scientific™ Culti-Loops™ API 20E Set contains quality control organisms supplied in Culti-Loops format.

Culti-Loops are ready to use, disposable inoculating loops containing stabilized, preserved, viable microorganisms. The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media. Culti-Loops are recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.

Includes:

Enterobacter cloacae ATCC™ 13047™†

Escherichia coli ATCC™ 25922™†

Klebsiella pneumoniae subsp. pneumoniae ATCC™ 35657™†

Proteus mirabilis ATCC™ 35659™†

Stenotrophomonas maltophilia ATCC™ 51331™†

Ordering Alerts:

Individually sealed in a foil pouch and packaged five loops per package.

TRUSTED_SUSTAINABILITY

Specifications

ATCC® 13047TM*
Description API 20E QC Set/5
Format Culti-Loops
Product Type Culti-Loops QC Set
Target Organism Class Organism Sets
Test Type Qualitative QC
Quantity 5 Loops/Organism

The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.