Learn More
Thermo Scientific™ Culti-Loops™ Aspergillus flavus ATCC™ 204304™
Shop All Thermo Scientific ProductsDescription
Confidently carry out testing of media, reagents, and identification & antimicrobial susceptibility kits and evaluate bacteriological procedures with ready-to-use Thermo Scientific™ Culti-Loops™ QC organisms.
Aspergillus flavus ATCC™ 204304™ is recommended to be used for antifungal susceptibility testing quality control. The strain is recommended for antifungal MIC applications QC testing by CLSI and EUCAST.
The loop may be dissolved in a liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000. Alternatively, it can be streaked directly onto appropriate media.
Includes:
Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
Specifications
Specifications
| ATCC® | ATCC™ 204304™ |
| Description | Aspergillus flavus ATCC™ 204304™ |
| Format | Culti-Loops™ |
| Target Organism Class | Fungi |
| Test Type | Qualitative QC |
| Quantity | 5 Loops/Pk. |
| Strain Designations | MCV-C#1 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.