Learn More
Thermo Scientific™ Culti-Loops™ Bacillus megaterium ATCC™ 14581™
Shop All Thermo Scientific ProductsDescription
Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media.
Ordering Information:
Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
Specifications
Specifications
| ATCC® | 14581TM* |
| Description | Bacillus megaterium |
| Product Type | Culti-Loops Quality Control Organism |
| Target Organism Class | Gram-positive Rods |
| Test Type | Qualitative QC |
| Packaging Type | 5 Loops/Pack |
| Quantity | 5 Loops |
| Strain Designations | WDCM00004, [BCRC 10608, CCM 2007, CCUG 1817, CIP 66.20, DSM 32, HAMBI 2018, IAM 13418, JCM 2506, KCTC 3007, LMG 7127, NBRC 15308, NCCB 75016, NCIMB 9376, NCTC 10342, NRIC 1710, NRRL B-14308, VKM B-512] |
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.