Learn More
Thermo Scientific™ Culti-Loops™ Enterobacter aerogenes ATCC™ 35028™
Catalog No. R4607081 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 Loop/Pk.
Catalog No. Quantity
R4607081 1 Loop/Pk.
1 options
This item is not returnable. View return policy
Catalog No. R4607081 Supplier Thermo Scientific™ Supplier No. R4607081
 Restricted Item
Add to Cart
Add to Cart
This item is not returnable. View return policy
Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media.
Ordering Information:
Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
TRUSTED_SUSTAINABILITY
Specifications
ATCC® 35028TM*
Description Enterobacter aerogenes ATCC™ 35028™
Product Type Culti-Loops Quality Control Organism
Target Organism Class Gram-negative Rods
Test Type Qualitative QC
Packaging Type 1 Loop/Pack
Quantity 1 Loop/Pk.
Strain Designations 413 [CDC N1001]
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.