Learn More
Thermo Scientific™ Culti-Loops™ Enterococcus faecalis ATCC™ 33186™
Shop All Thermo Scientific ProductsDescription
Confidently carry out testing of media, reagents, and identification & antimicrobial susceptibility kits and evaluate bacteriological procedures with ready-to-use Thermo Scientific™ Culti-Loops™ QC organisms.
Enterococcus faecalis™ ATCC™ 33186™ is recommended to be used to assess suitability of Mueller Hinton Agar or broth for sulfonamide and trimethoprim MIC & disc diffusion tests (eg. trimethoprim-sulfamethoxazole discs).
Enterococcus faecalis™ ATCC™ 33186™ can be used to assess Mueller-Hinton medium suitability for sulfonamide and trimethoprim testing.
Mueller-Hinton agar or broth medium containing excessive amounts of thymidine or thymine can reverse the inhibitory effect of sulfonamides and trimethoprim, which may result in smaller and less distinct disc diffusion zones and false-resistance reports.
With broth microdilution MIC testing, suitability of Mueller-Hinton broth for sulfonamide & trimethoprim tests can be evaluated by reviewing ease of reading the results: the end points should be clear at 80% or greater reduction in growth compared to the control.
Technique:
The loop can be streaked directly onto appropriate media.
Alternatively, it may be dissolved in a liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000.
Includes:
Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
Specifications
Specifications
| ATCC® | ATCC™ 33186™ |
| Description | Enterococcus faecalis™ ATCC™ 33186™ |
| Format | Culti-Loops |
| Target Organism Class | Gram-positive Cocci |
| Test Type | Qualitative QC |
| Quantity | 5 Loops/Pk. |
| Strain Designations | CN 478 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.