Learn More
Description
Confidently carry out testing of media, reagents, and identification & antimicrobial susceptibility kits and evaluate bacteriological procedures with ready-to-use Thermo Scientific™ Culti-Loops™ QC organisms.
Escherichia coli NCTC 13353™ is an extended spectrum β-lactamase (ESBL) producer expressing Cefotaximase-Munich 15 (CTX-M-15) ESBL.
The Escherichia coli NCTC 13353 strain is recommended to be used with antimicrobial susceptibility quality control testing, specifically with β-lactam combination agent QC testing for disc diffusion and MIC tests.
The strain is resistant to cephalosporins and has CLSI defined QC ranges for various cephalosporins and cephalosporin/ β-lactamase inhibitor combination agents.
Technique:
The loop can be streaked directly onto appropriate media.
Alternatively, it may be dissolved in a liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000.
Includes:
Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
Specifications
Specifications
| Description | Escherichia coli™ NCTC 13353™ |
| Format | Culti-Loops™ |
| Target Organism Class | Gram-negative Rods |
| Test Type | Qualitative QC |
| NCTC® | NCTC 13353™ |
| Quantity | 5 Loops/Pk. |
| Strain Designations | E0487 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.