Learn More
Description
Confidently carry out testing of media, reagents, and identification & antimicrobial susceptibility kits and evaluate bacteriological procedures with ready-to-use Thermo Scientific™ Culti-Loops™ QC organisms.
Escherichia coli NCTC 13846™ is recommended to be used for antimicrobial susceptibility quality control testing.
Escherichia coli NCTC 13846™ is colistin resistant conferred by mcr-1 gene. EUCAST recommends the strain to be tested as part of routine QC for colistin MIC testing.
Technique:
The loop can be streaked directly onto appropriate media.
Alternatively, it may be dissolved in a liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000.
Includes:
Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
Specifications
Specifications
| Description | Escherichia coli™ NCTC 13846™ |
| Format | Culti-Loops™ |
| Target Organism Class | Gram-negative Rods |
| Test Type | Qualitative QC |
| NCTC® | NCTC 13846™ |
| Quantity | 5 Loops/Pk. |
| Strain Designations | DSM 105182 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.