Learn More
Thermo Scientific™ Culti-Loops™ Kirby Bauer Susceptibility QC Set
Shop All Thermo Scientific ProductsDescription
Thermo Scientific™ Culti-Loops™ Kirby Bauer Susceptibility Set contains quality control organisms supplied in Culti-Loops format.
Culti-Loops are ready to use, disposable inoculating loops containing stabilized, preserved, viable microorganisms. The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media. Culti-Loops are recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures
.Includes:
Escherichia coli ATCC™ 25922™†
Escherichia coli ATCC™ 35218™†
Pseudomonas aeruginosa ATCC™ 27853™†
Staphylococcus aureus supsp. aureus ATCC™ 25923™†
Ordering Alerts:
Individually sealed in a foil pouch and packaged five loops per package.
Specifications
Specifications
| ATCC® | 25922TM* |
| Description | Kirby Bauer Susceptibility QC Set/4 |
| Format | Culti-Loops |
| Product Type | Culti-Loop |
| Target Organism Class | Organism Sets |
| Test Type | Qualitative QC |
| Quantity | 5 Loops/Organism |
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.