Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Culti-Loops™ Klebsiella pneumoniae ATCC™ BAA-2814™

Catalog No. R4601316 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 Loops/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R4601316 5 Loops/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. R4601316 Supplier Thermo Scientific™ Supplier No. R4601316
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Perform quality control testing and performance evaluation of microbiological procedures with these ready-to-use bacteriological loops.

Confidently carry out testing of media, reagents, and identification & antimicrobial susceptibility kits and evaluate bacteriological procedures with ready-to-use Thermo Scientific™ Culti-Loops™ QC organisms.

Klebsiella pneumoniae ATCC™ BAA-2814™ is recommended to be used for antimicrobial susceptibility testing quality control with disc diffusion and MIC applications.

The Klebsiella pneumoniae ATCC™ BAA-2814™ strain produces Klebsiella pneumoniae carbapenemase KPC-3 and β-lactamase enzymes SHV-11 & TEM-1. The strain is recommended for β-lactam combination agent QC testing, such as Meropenem-vaborbactam and Imipenem-relebactam, by CLSI and EUCAST.

Technique:

The loop can be streaked directly onto appropriate media.

Alternatively, it may be dissolved in a liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000.

Includes:

Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.

TRUSTED_SUSTAINABILITY

Specifications

ATCC® ATCC™ BAA-2814™
Description Klebsiella pneumoniae ATCC™ BAA-2814™
Format Culti-Loops™
Target Organism Class Gram-negative Rods
Test Type Qualitative QC
Quantity 5 Loops/Pk.
Strain Designations KP1074 [B21]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.