Learn More
Description
Confidently carry out testing of media, reagents, and identification & antimicrobial susceptibility kits and evaluate bacteriological procedures with ready-to-use Thermo Scientific™ Culti-Loops™ QC organisms.
Legionella anisa NCTC 11974™ is recommended as a quality control strain for culture media in water quality testing according to ISO 11731:2016.
The loop can be streaked directly onto appropriate media.
Alternatively, it may be dissolved in a liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000.
Includes:
Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
Specifications
Specifications
| ATCC® | NCTC 11974™ |
| Description | Legionella anisa NCTC 11974™ |
| Format | Culti-Loops™ |
| Target Organism Class | Gram-negative Bacilli |
| Test Type | Qualitative QC |
| Quantity | 5 Loops/Pk. |
| Strain Designations | ATCC 35292, CCUG 29665, CIP 103870, DSM 17627, WDCM 00106 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.