Learn More
Thermo Scientific™ Culti-Loops™ Microbacterium liquefaciens ATCC™ BAA-1819™
Shop All Thermo Scientific ProductsDescription
Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media.
Ordering Information:
Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
Specifications
Specifications
| ATCC® | BAA-1819TM* |
| Certifications/Compliance | Culti-Loops are CE Marked. |
| Description | Microbacterium liquefaciens |
| Detects | Microbacterium liquefaciens ATCC™ BAA-1819™ |
| Format | Culti-Loops |
| Product Type | Culti-Loops Quality Control Organism |
| Target Organism Class | Gram-negative Rods |
| Test Type | Qualitative QC |
| Packaging Type | 5 Loops/Pack |
| Quantity | 5 Loops/Pk. |
| Show More |
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.