Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Culti-Loops™ Mycobacterium fortuitum subsp. fortuitum ATCC™ 6841™

Catalog No. R4610012 Shop All Thermo Scientific Products
Change view
Click to view available options
Description:
Mycobacterium fortuitum subsp. fortuitum ATCC™ 6841™
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
R4610012 Mycobacterium fortuitum subsp. fortuitum ATCC™ 6841™
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. R4610012 Supplier Thermo Scientific™ Supplier No. R4610012
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.

Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.

The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media.

Ordering Information:

Culti-Loops are individually sealed in a foil pouch and packaged one loop per package.

TRUSTED_SUSTAINABILITY

Specifications

ATCC® 6841TM*
Description Mycobacterium fortuitum subsp. fortuitum ATCC™ 6841™
Format Culti-Loops
Product Type Mycobacti-Loop
Target Organism Class Fastidious
Packaging Type 1 Loop/Pack
Strain Designations WDCM00072, [TMC 1529]

*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.