Learn More
Description
Confidently carry out testing of media, reagents, and identification & antimicrobial susceptibility kits and evaluate the bacteriological procedures with Thermo Scientific™ Culti-Loops™ ready-to-use QC organisms.
Proteus mirabilis 511-116 strain produces extended-spectrum β-lactamase (ESBL). This strain is recommended to be used as a positive control strain with Thermo Scientific™ Spectra™ ESBL Medium following the HAI surveillance programs.
The loop can be streaked directly onto appropriate media. Alternatively, it may be dissolved in a liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000.
Ordering Information:
Culti-Loops are individually sealed in a foil pouch and packaged five loops per package
Specifications
Specifications
| Format | Culti-Loops |
| Product Type | Culti-Loops Quality Control Organism |
| Species | Proteus species |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.