Learn More
Thermo Scientific™ Culti-Loops™ Staphylococcus aureus NCTC 12493™
Shop All Thermo Scientific ProductsDescription
Confidently carry out testing of media, reagents, and identification & antimicrobial susceptibility kits and evaluate bacteriological procedures with ready-to-use Thermo Scientific™ Culti-Loops™ QC organisms.
Staphylococcus aureus NCTC 12493™ is methicillin resistant (MRSA) conferred by mec-A gene. EUCAST recommends this strain to be tested as part of extended QC for cefoxitin disc testing.
Technique:
The loop can be streaked directly onto appropriate media.
Alternatively, it may be dissolved in a liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000.
Includes:
Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
Specifications
Specifications
| Description | Staphylococcus aureus NCTC 12493™ |
| Format | Culti-Loops™ |
| Target Organism Class | Gram-positive Cocci |
| Test Type | Qualitative QC |
| NCTC® | NCTC 12493™ |
| Quantity | 5 Loops/Pk. |
| Strain Designations | MQCL 113, WDCM 00212 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.