Learn More
Takara Bio CULTILIFE CELL-CULTURE BAGS 10
SDP
Catalog No. NC1901171
Encompass
ultiLife cell culture bags provide a sterile, closed system for growing cells that reduces the risk of both culture contamination and user infection. CultiLife 215 cell culture bags are designed for medium to large-scale cell culture. Each bag has a culture area of 215 cm2.
Catalog No. NC1901171 Supplier Takara Bio Supplier No. FU0005
ultiLife cell culture bags provide a sterile, closed system for growing cells that reduces the risk of both culture contamination and user infection. CultiLife 215 cell culture bags are designed for medium to large-scale cell culture. Each bag has a culture area of 215 cm2.