Learn More
Thermo Scientific™ CultiMaxx™ Shelving System for G-Rex 500M-CS Bioreactors
GSA_VA
Catalog No. 11670000 Shop All Thermo Scientific Products
Change view
Click to view available options
No. of Shelves:
3
Material:
Stainless Steel (18/8)
Catalog No. No. of Shelves Material
11-670-000 3 Stainless Steel (18/8)
1 options
Catalog No. 11-670-000 Supplier Thermo Scientific™ Supplier No. 50164781
Add to Cart
Add to Cart
CultiMaxx™ Shelving Systems to support G-Rex 500M-CS Bioreactors
Maximize the capacity, increase throughput and optimize cell growth in your lab using the CultiMaxx shelving system in a range of large capacity CO2 Incubators.
Maximize incubator space  
  • These shelving systems are designed to support scale-out of the G-Rex 500M-CS* in cell therapy production.
  • With an in-depth review of the shelving design, this new system provides up to a 150% increase in capacity for the G-Rex, or another way to look at it: 2 1/2 incubators in 1.
 Easy installation
  • Shelving systems are retrofittable and can be self-installed.
*G-Rex is a registered trademark of Wilson Wolf Corporation. G-Rex bioreactors must be ordered from Wilson Wolf Corporation.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Description Shelving system to support up to 10 G-Rex 500M-CS bioreactors for Heracell VIOS 250i (CR), Forma Steri-Cycle i250 (CR) CO2 incubator
For Use With (Equipment) Heracell™ Vios™ 250i, Forma™ Steri-Cycle™ i250, Heracell™ Vios™ 250i CR, Forma™ Steri-Cycle™ i250 CR CO2 Incubators, Heracell Vios 250i, Forma Steri-Cycle i250 and CR/CTS series
Material Stainless Steel (18/8)
No. of Shelves 3
Type Shelving System
Includes 3 expanded shelves with reinforced design, shelf supports and shelf rails