Learn More
Sigma Aldrich Fine Chemicals Biosciences CultispherR -S standard po10G
Catalog No. 502904804
Encompass
Macroporous gelatin-coated microcarrier beads. General purpose beads for anchorage-dependent cells.
Catalog No. 50-290-4804 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. M904310G
Macroporous gelatin-coated microcarrier beads. General purpose beads for anchorage-dependent cells.