Learn More
Electron Microscopy Sciences CULTRE TBE PS 5MM STRL 25 SEAL
SDP
Catalog No. 502413082
Encompass_Preferred
Disposable sterile tubes with caps made of polyethylene (PE) and have the popular two position design (up/loose for aerobic work and down/sealed for anaerobic cultures). Polystyrene tubes may be centrifuged at up to 2000xg. Polypropylene tubes are able to withstand higher centrifugal forces, up to 4000xg.
Catalog No. 50-241-3082 Supplier Electron Microscopy Sciences Supplier No. 6463908
Disposable sterile tubes with caps made of polyethylene (PE) and have the popular two position design (up/loose for aerobic work and down/sealed for anaerobic cultures). Polystyrene tubes may be centrifuged at up to 2000xg. Polypropylene tubes are able to withstand higher centrifugal forces, up to 4000xg.