Learn More
Alkali Scientific Culture Tubes, 5mL, 12 x 75mm Polypropylene (PP) with Cap, Sterile, 25/Bag, 1000/cs
SDP
Catalog No. 502336031
Encompass_Preferred
These disposable sterile tubes are made of high quality polypropylene (PP) that will withstand temperatures from -190˚ to +120˚C. They are also available in crystal clear polystyrene (PS) with a more limited range of 0˚ to 70˚C. Caps are made of polyethylene (PE) and have the popular two position design (up/loose for aerobic work and down/sealed for anaerobic cultures). Polystyrene tubes may be centrifuged at up to 2000xg. Polypropylene tubes withstand higher forces, up to 4000xg.
Catalog No. 50-233-6031 Supplier Alkali Scientific Supplier No. CT5301
These disposable sterile tubes are made of high quality polypropylene (PP) that will withstand temperatures from -190˚ to +120˚C. They are also available in crystal clear polystyrene (PS) with a more limited range of 0˚ to 70˚C. Caps are made of polyethylene (PE) and have the popular two position design (up/loose for aerobic work and down/sealed for anaerobic cultures). Polystyrene tubes may be centrifuged at up to 2000xg. Polypropylene tubes withstand higher forces, up to 4000xg.