Learn More
R&D Systems™ Cultrex Rat Collagen I, 5 mg
Specifications
Specifications
| Gene ID (Entrez) | 1277 |
| Quantity | 5 mg |
| Storage Requirements | Product is stable for 3 months from the date of receipt when stored at 2 - 8°C. Keep sterile. |
| Gene Alias | Collagen 1, Collagen I |
| Species | Rat |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.