Learn More
R&D Systems™ Cultrex™ Stem Cell Qualified Human Fibronectin PathClear™
SDP
Catalog No. 342000103
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
34-200-0103 1 mg
1 options
This item is not returnable. View return policy
Catalog No. 34-200-0103 Supplier R&D Systems™ Supplier No. 342000103
Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.
This item is not returnable. View return policy
R&D Systems™ Cultrex™ Stem Cell Qualified Human Fibronectin PathClear™ provides a functionally defined and effective feeder-free surface for the attachment and maintenance of embryonic stem cells in a pluripotent state.
Specifications
Quantity 1 mg
Source Human plasma