Learn More
Cole-Parmer Techne 6027689 Glass Cell Culture Flask, 250 mL; 1/ea
Supplier:  Cole-Parmer 2183734
Encompass_Preferred
Culture vessels come calibrated and have two side ports and a central filling port. All culture vessel parts can be autoclaved at 121°C and 15 psi for 30 minutes. Completely seal vessels for use with pathogenic materials. Silicone-encapsulated stir rods reduce the possibility of cells attaching to and growing on the surface. Order a complete culture vessel (flask, stir rod, and caps) to use with these biological stirrers or order just a flask to use with an existing stirrer.
Catalog No. 50-196-1003