Learn More
Ibidi USA Culture-Insert 4 Well in µ-Dish 35mm, High, ibiTreat - Tissue Culture Treated Polymer Coverslip
SDP
Catalog No. 501149018
Encompass
The ibidi Culture-Insert family is developed for wound healing, co-cultivation, and migration assays of adherent cells. A proprietary biocompatible adhesive is applied to the underside of the Culture-Insert which allows it to adhere to any flat, clean, and dry surface. The Culture-Insert 4 Well consists of four wells, representing the four quarters of the round Culture-Insert. The wells are separated by a wall of 500 µm. When the wells are filled with adherent cells, a cell–free gap of 500 µm is created between the adjacent wells after removing the Culture-Insert 4 Well. Additionally, a center gap of 1000 µm is created, allowing to observe the migration of all cell types at once. Live-cell imaging can be used to analyze the gap closures. The Culture-Insert 4 Well comes ready-to-use in our µ-Dish 35 mm, High, ibiTreat dishes. Sterile, 30 pcs./box.
This item is not returnable. View return policy
Catalog No. 50-114-9018 Supplier Ibidi USA Supplier No. 80466
The ibidi Culture-Insert family is developed for wound healing, co-cultivation, and migration assays of adherent cells. A proprietary biocompatible adhesive is applied to the underside of the Culture-Insert which allows it to adhere to any flat, clean, and dry surface. The Culture-Insert 4 Well consists of four wells, representing the four quarters of the round Culture-Insert. The wells are separated by a wall of 500 µm. When the wells are filled with adherent cells, a cell–free gap of 500 µm is created between the adjacent wells after removing the Culture-Insert 4 Well. Additionally, a center gap of 1000 µm is created, allowing to observe the migration of all cell types at once. Live-cell imaging can be used to analyze the gap closures. The Culture-Insert 4 Well comes ready-to-use in our µ-Dish 35 mm, High, ibiTreat dishes. Sterile, 30 pcs./box.