Learn More
Lonza Walkersville TheraPEAK MSCBM Medium 1L
Catalog No. 5000496277
Encompass
Serum-free media such as TheraPEAK MSCGM Mesenchymal Stem Cell Medium are intended to support cell growth without serum supplementation Lonza s TheraPEAK MSCGM Mesenchymal Stem Cell Medium is of primary interest to those seeking maximal control over research and manufacturing variables TheraPEAK MSCGM Mesenchymal Stem Cell Medium does not contain phenol red or antibiotics
Catalog No. 50-004-96277 Supplier Lonza Walkersville Supplier No. BEBP12934Q
Serum-free media such as TheraPEAK MSCGM Mesenchymal Stem Cell Medium are intended to support cell growth without serum supplementation Lonza s TheraPEAK MSCGM Mesenchymal Stem Cell Medium is of primary interest to those seeking maximal control over research and manufacturing variables TheraPEAK MSCGM Mesenchymal Stem Cell Medium does not contain phenol red or antibiotics