Learn More
Lonza Walkersville TheraPEAK PBS w/o Ca Mg GMP
Catalog No. 5000496458
Encompass
TheraPEAK PBS serves as a wash solution in a wide variety of laboratory applications this GMP produced version fits excellent with downstream processing in cell and gene therapy and protein manufacturing applications This BioWhittaker Cell Culture Media is produced according to cGMP and labelled For Further Manufacturing (FFM) GMP Products are intended for research or further manufacturing use only THIS PRODUCT IS NOT INTENDED FOR DIRECT THERAPEUTIC USE IN HUMANS
Catalog No. 50-004-96458 Supplier Lonza Walkersville Supplier No. BEBP17516Q
Add to Cart
Add to Cart
TheraPEAK PBS serves as a wash solution in a wide variety of laboratory applications this GMP produced version fits excellent with downstream processing in cell and gene therapy and protein manufacturing applications This BioWhittaker Cell Culture Media is produced according to cGMP and labelled For Further Manufacturing (FFM) GMP Products are intended for research or further manufacturing use only THIS PRODUCT IS NOT INTENDED FOR DIRECT THERAPEUTIC USE IN HUMANS