Learn More
Lonza Walkersville DPBS Powder 10L
Catalog No. 5000496409
Encompass
Lonza s 15-512 powder is a skeleton DPBS powder without calcium and magnesium to reconstitute all liquid DPBS formulations This DPBS powder without calcium and magnesium is available in 10 50 and 1000 containers This means these containers can make 10 50 or 1000L of liquid medium when dissolved in water This BioWhittaker Cell Culture Powder is produced according to cGMP and labelled For Further Manufacturing (FFM) GMP Products are intended for research or further manufacturing use only THIS PRODUCT IS NOT INTENDED FOR DIRECT THERAPEUTIC USE IN HUMANS
Catalog No. 50-004-96409 Supplier Lonza Walkersville Supplier No. BE15512D
Lonza s 15-512 powder is a skeleton DPBS powder without calcium and magnesium to reconstitute all liquid DPBS formulations This DPBS powder without calcium and magnesium is available in 10 50 and 1000 containers This means these containers can make 10 50 or 1000L of liquid medium when dissolved in water This BioWhittaker Cell Culture Powder is produced according to cGMP and labelled For Further Manufacturing (FFM) GMP Products are intended for research or further manufacturing use only THIS PRODUCT IS NOT INTENDED FOR DIRECT THERAPEUTIC USE IN HUMANS