Learn More
Lonza Walkersville L-Glutamine GMP 200mM 100ml
Catalog No. 5000496253
Encompass
L-Glutamine GMP supplied as 29 3 mg/ml in 0 85% NaCL 200 mM for application where L-Glutamine GMP is required This BioWhittaker Cell Culture Media is produced according to cGMP and labelled For Further Manufacturing (FFM) GMP Products are intended for research or further manufacturing use only The products confirm with cGMP manufacturing compliance expectations for medical devices including 21 CFR Part 820 and ISO 13485 THIS PRODUCT IS NOT INTENDED FOR DIRECT THERAPEUTIC USE IN HUMANS
Catalog No. 50-004-96253 Supplier Lonza Walkersville Supplier No. BEBP17605E
L-Glutamine GMP supplied as 29 3 mg/ml in 0 85% NaCL 200 mM for application where L-Glutamine GMP is required This BioWhittaker Cell Culture Media is produced according to cGMP and labelled For Further Manufacturing (FFM) GMP Products are intended for research or further manufacturing use only The products confirm with cGMP manufacturing compliance expectations for medical devices including 21 CFR Part 820 and ISO 13485 THIS PRODUCT IS NOT INTENDED FOR DIRECT THERAPEUTIC USE IN HUMANS