Learn More
Lonza Walkersville Water for Cell Culture 20L
Catalog No. 5000496467
Encompass
Bag filled with Water for Cell Culture Water For Injection (WFI) quality water is prepared by ultrafiltration reveresed osmosis deionisaton and sterile filtration Lonza s Water for Cell Culture is aseptically processed from a validated Water for Injection (WFI) system that meets current USP specifications This BioWhittaker Cell Culture Media is produced according to cGMP and labelled For Further Manufacturing (FFM) GMP Products are intended for research or further manufacturing use only THIS PRODUCT IS NOT INTENDED FOR DIRECT THERAPEUTIC USE IN HUMANS
Catalog No. 50-004-96467 Supplier Lonza Walkersville Supplier No. BE02038P20
Add to Cart
Add to Cart
Bag filled with Water for Cell Culture Water For Injection (WFI) quality water is prepared by ultrafiltration reveresed osmosis deionisaton and sterile filtration Lonza s Water for Cell Culture is aseptically processed from a validated Water for Injection (WFI) system that meets current USP specifications This BioWhittaker Cell Culture Media is produced according to cGMP and labelled For Further Manufacturing (FFM) GMP Products are intended for research or further manufacturing use only THIS PRODUCT IS NOT INTENDED FOR DIRECT THERAPEUTIC USE IN HUMANS