Learn More
Lonza Walkersville MEM Eagle Vitamin Mix GMP 100x
Catalog No. 5000496226
Encompass
MEM Eagle Vitamin Mixture GMP is used to support cell growth in tissue culture media Usage 10 mL per liter Note differences in appearance in the frozen state do not affect performance of the stock This BioWhittaker Cell Culture Media is produced according to cGMP and labelled For Further Manufacturing (FFM) GMP Products are intended for research or further manufacturing use only The products confirm with cGMP manufacturing compliance expectations for medical devices including 21 CFR Part 820 and ISO 13485 THIS PRODUCT IS NOT INTENDED FOR DIRECT THERAPEUTIC USE IN HUMANS
Catalog No. 50-004-96226 Supplier Lonza Walkersville Supplier No. BEBP13607C
MEM Eagle Vitamin Mixture GMP is used to support cell growth in tissue culture media Usage 10 mL per liter Note differences in appearance in the frozen state do not affect performance of the stock This BioWhittaker Cell Culture Media is produced according to cGMP and labelled For Further Manufacturing (FFM) GMP Products are intended for research or further manufacturing use only The products confirm with cGMP manufacturing compliance expectations for medical devices including 21 CFR Part 820 and ISO 13485 THIS PRODUCT IS NOT INTENDED FOR DIRECT THERAPEUTIC USE IN HUMANS