Learn More
Lonza Walkersville PowerFeed A - CD Medium 1L
Catalog No. 5000496439
Encompass
PowerFeed A Chemically Defined and Protein-free Liquid Medium is also non-animal origin and a liquid feed The feed is formulated to maximize cell growth using Lonza CHO cell culture media ProCHO 4 -5 and PowerCHO 2 This BioWhittaker Cell Culture Media is produced according to cGMP and labelled For Further Manufacturing (FFM) GMP Products are intended for research or further manufacturing use only THIS PRODUCT IS NOT INTENDED FOR DIRECT THERAPEUTIC USE IN HUMANS
Catalog No. 50-004-96439 Supplier Lonza Walkersville Supplier No. BE02044Q
PowerFeed A Chemically Defined and Protein-free Liquid Medium is also non-animal origin and a liquid feed The feed is formulated to maximize cell growth using Lonza CHO cell culture media ProCHO 4 -5 and PowerCHO 2 This BioWhittaker Cell Culture Media is produced according to cGMP and labelled For Further Manufacturing (FFM) GMP Products are intended for research or further manufacturing use only THIS PRODUCT IS NOT INTENDED FOR DIRECT THERAPEUTIC USE IN HUMANS