Learn More
Lonza Walkersville XTREME CHO Feed CD Liquid 1L
Catalog No. 5000496285
Encompass
XTREME CHO Feed Liquid is a chemically defined protein-free and non-animal origin liquid feed The feed is formulated to maximize cell growth using Lonza CHO cell culture media PowerCHO 2 The XTREME CHO Feed has low salts for better osmolarity control This BioWhittaker Cell Culture Media is produced according to cGMP and labelled For Further Manufacturing (FFM) GMP Products are intended for research or further manufacturing use only THIS PRODUCT IS NOT INTENDED FOR DIRECT THERAPEUTIC USE IN HUMANS
Catalog No. 50-004-96285 Supplier Lonza Walkersville Supplier No. BE02056Q
XTREME CHO Feed Liquid is a chemically defined protein-free and non-animal origin liquid feed The feed is formulated to maximize cell growth using Lonza CHO cell culture media PowerCHO 2 The XTREME CHO Feed has low salts for better osmolarity control This BioWhittaker Cell Culture Media is produced according to cGMP and labelled For Further Manufacturing (FFM) GMP Products are intended for research or further manufacturing use only THIS PRODUCT IS NOT INTENDED FOR DIRECT THERAPEUTIC USE IN HUMANS