Learn More
Lonza Walkersville TheraPEAK X-VIVO-15 w/rec tran
Catalog No. 5000496359
Encompass
TheraPEAK X-VIVO 15 Media is a high performing media that provides nutritionally adequate and balanced environments for a variety of hematopoietic cells including T cells Natural killer cells (NK) Peripheral blood lymphocytes (PBL) Tumor infiltrating lymphocytes (TIL) Dendritic cells (DC) Human monocytes Macrophage cells TheraPEAK X-VIVO 15 Media are mostly known for its ability to support the proliferation of purified CD3 cells isolated from peripheral blood and human tumors making it well suited for CAR-T applications TheraPEAK X-VIVO 15 Media are similar in composition to X-VIVO 10 Media but is better suited for rapidly growing cells specifically cells of the immune system All TheraPEAK Products are produced according to applicable GMP standards and follow the USP/EP guidance for cell and gene therapy raw materials
Catalog No. 50-004-96359 Supplier Lonza Walkersville Supplier No. BEBP02054P1
TheraPEAK X-VIVO 15 Media is a high performing media that provides nutritionally adequate and balanced environments for a variety of hematopoietic cells including T cells Natural killer cells (NK) Peripheral blood lymphocytes (PBL) Tumor infiltrating lymphocytes (TIL) Dendritic cells (DC) Human monocytes Macrophage cells TheraPEAK X-VIVO 15 Media are mostly known for its ability to support the proliferation of purified CD3 cells isolated from peripheral blood and human tumors making it well suited for CAR-T applications TheraPEAK X-VIVO 15 Media are similar in composition to X-VIVO 10 Media but is better suited for rapidly growing cells specifically cells of the immune system All TheraPEAK Products are produced according to applicable GMP standards and follow the USP/EP guidance for cell and gene therapy raw materials