Learn More
Lonza Walkersville TheraPRO CHO Expansion Med 1L
Catalog No. 5000496446
Encompass
TheraPRO CHO Expansion Medium supports the growth and viability of GS-CHO cells from revival subculture and transfection prior to scale up into production Establishing a viable starting culture of CHO cells is essential for a successful production run The TheraPRO CHO Expansion Medium is chemically defined and optimized to be used with the TheraPRO CHO Production Medium and Feed to ensure a seamless transition from revival to production preparation All components of the TheraPRO CHO Media System are manufactured in ISO 13485 certified facilities in accordance with GMP and are accompanied by a Drug Master File (DMF) and assistance with regulatory submissions
Catalog No. 50-004-96446 Supplier Lonza Walkersville Supplier No. BESP1630Q
TheraPRO CHO Expansion Medium supports the growth and viability of GS-CHO cells from revival subculture and transfection prior to scale up into production Establishing a viable starting culture of CHO cells is essential for a successful production run The TheraPRO CHO Expansion Medium is chemically defined and optimized to be used with the TheraPRO CHO Production Medium and Feed to ensure a seamless transition from revival to production preparation All components of the TheraPRO CHO Media System are manufactured in ISO 13485 certified facilities in accordance with GMP and are accompanied by a Drug Master File (DMF) and assistance with regulatory submissions