Learn More
Lonza Walkersville Pro293s SF Medium in Suspens
Catalog No. 5000496295
Encompass
Pro293 chemically defined serum-free media were optimized to support high-density growth and recombinant protein production in 293 neonatal kidney cells They are chemically defined to ease regulatory compliance and downstream protein purification They contain very low levels of recombinant human insulin and are free of animal-origin components Pro293s Medium mdash for 293 cells growing in suspension culture Pro293a Medium mdash for 293 cells growing in adherent culture Both media do not contain L-glutamine or phenol red Pluronic F-68 is present at 0 1% This BioWhittaker Cell Culture Media is produced according to cGMP and labelled For Further Manufacturing (FFM)
Catalog No. 50-004-96295 Supplier Lonza Walkersville Supplier No. BEBP02025Q
Pro293 chemically defined serum-free media were optimized to support high-density growth and recombinant protein production in 293 neonatal kidney cells They are chemically defined to ease regulatory compliance and downstream protein purification They contain very low levels of recombinant human insulin and are free of animal-origin components Pro293s Medium mdash for 293 cells growing in suspension culture Pro293a Medium mdash for 293 cells growing in adherent culture Both media do not contain L-glutamine or phenol red Pluronic F-68 is present at 0 1% This BioWhittaker Cell Culture Media is produced according to cGMP and labelled For Further Manufacturing (FFM)