Learn More
Lonza Walkersville TheraPEAK MSCGM BulletKit
Catalog No. 5000496280
Encompass
The medium has been specifically designed and tested to ensure that the medium neither promotes spontaneous differentiation of cells nor inhibits cell differentiation to osteogenic chondrogenic or adipogenic lineages (when properly stimulated with an appropriate differentiation media) Serum-free media such as TheraPEAK MSCGM Mesenchymal Stem Cell Growth Medium are intended to support cell growth without serum supplementation Lonza s TheraPEAK MSCGM Mesenchymal Stem Cell Growth Medium is of primary interest to those seeking maximal control over research and manufacturing variables TheraPEAK MSCGM Mesenchymal Stem Cell Growth Medium does not contain phenol red or antibiotics
Catalog No. 50-004-96280 Supplier Lonza Walkersville Supplier No. BEBP18936
The medium has been specifically designed and tested to ensure that the medium neither promotes spontaneous differentiation of cells nor inhibits cell differentiation to osteogenic chondrogenic or adipogenic lineages (when properly stimulated with an appropriate differentiation media) Serum-free media such as TheraPEAK MSCGM Mesenchymal Stem Cell Growth Medium are intended to support cell growth without serum supplementation Lonza s TheraPEAK MSCGM Mesenchymal Stem Cell Growth Medium is of primary interest to those seeking maximal control over research and manufacturing variables TheraPEAK MSCGM Mesenchymal Stem Cell Growth Medium does not contain phenol red or antibiotics