Learn More
Lonza Walkersville PowerCHO 2 SF Medium 10L bag
Catalog No. 5000496271
Encompass
PowerCHO Media are hydrolysate-free serum-free and non-animal origin media for supporting high-density CHO cells in suspension For therapeutic bioprocessing applications these protein-free formulations also facilitate both downstream purification and regulatory compliance PowerCHO contains HEPES and Pluronic F-68 but does not contain L-glutamine phenol red hypoxanthine and thymidine PowerCHO 2 Chemically Defined Serum-free CHO Media is optimized for both cell growth and protein production The 10 L bag provides a flexible single use approach This BioWhittaker Cell Culture Media is produced according to cGMP and labelled For Further Manufacturing (FFM) GMP Products are intended for research or further manufacturing use only THIS PRODUCT IS NOT INTENDED FOR DIRECT THERAPEUTIC USE IN HUMANS
Catalog No. 50-004-96271 Supplier Lonza Walkersville Supplier No. BE12771P10
PowerCHO Media are hydrolysate-free serum-free and non-animal origin media for supporting high-density CHO cells in suspension For therapeutic bioprocessing applications these protein-free formulations also facilitate both downstream purification and regulatory compliance PowerCHO contains HEPES and Pluronic F-68 but does not contain L-glutamine phenol red hypoxanthine and thymidine PowerCHO 2 Chemically Defined Serum-free CHO Media is optimized for both cell growth and protein production The 10 L bag provides a flexible single use approach This BioWhittaker Cell Culture Media is produced according to cGMP and labelled For Further Manufacturing (FFM) GMP Products are intended for research or further manufacturing use only THIS PRODUCT IS NOT INTENDED FOR DIRECT THERAPEUTIC USE IN HUMANS