Learn More
Lonza Walkersville PowerCHO 2 Powder Kit 100L
Catalog No. 5000496225
Encompass
PowerCHO 2 Powder kit 100 L is a hydrolysate-free serum-free and non-animal origin medium for supporting high-density CHO cells in suspension For therapeutic bioprocessing applications these protein-free formulations also facilitate both downstream purification and regulatory compliance PowerCHO 2 Powder kit 100 L contains HEPES and Pluronic F-68 but does not contain L-glutamine phenol red hypoxanthine and thymidine PowerCHO 2 Powder kit 100 L is a Chemically Defined Serum-free CHO Medium optimized for both cell growth and protein production The powder kit contains several components to produce the liquid medium PowerCHO 2 Powder kit 100 L is compatible with fed-batch cultiring by using one of feeds eCHO Feed Power Feed A CHO Xtreme Feed This BioWhittaker Cell Culture Media is produced according to cGMP and labelled For Further Manufacturing (FFM) GMP Products are intended for research or further manufacturing use only
Catalog No. 50-004-96225 Supplier Lonza Walkersville Supplier No. BE15771NJ
PowerCHO 2 Powder kit 100 L is a hydrolysate-free serum-free and non-animal origin medium for supporting high-density CHO cells in suspension For therapeutic bioprocessing applications these protein-free formulations also facilitate both downstream purification and regulatory compliance PowerCHO 2 Powder kit 100 L contains HEPES and Pluronic F-68 but does not contain L-glutamine phenol red hypoxanthine and thymidine PowerCHO 2 Powder kit 100 L is a Chemically Defined Serum-free CHO Medium optimized for both cell growth and protein production The powder kit contains several components to produce the liquid medium PowerCHO 2 Powder kit 100 L is compatible with fed-batch cultiring by using one of feeds eCHO Feed Power Feed A CHO Xtreme Feed This BioWhittaker Cell Culture Media is produced according to cGMP and labelled For Further Manufacturing (FFM) GMP Products are intended for research or further manufacturing use only