Learn More
Lonza Walkersville Polyethylene Glycol for 100L
Catalog No. 5000496205
Encompass
PEG for PowerCHO 2 Powder Kit is a component of PowerCHO 2 Powder 100 L Kit (sold separately or as part of the kit) This BioWhittaker Cell Culture Media is produced according to cGMP and labelled For Further Manufacturing (FFM) GMP Products are intended for research or further manufacturing use only THIS PRODUCT IS NOT INTENDED FOR DIRECT THERAPEUTIC USE IN HUMANS
Catalog No. 50-004-96205 Supplier Lonza Walkersville Supplier No. VPW070J
PEG for PowerCHO 2 Powder Kit is a component of PowerCHO 2 Powder 100 L Kit (sold separately or as part of the kit) This BioWhittaker Cell Culture Media is produced according to cGMP and labelled For Further Manufacturing (FFM) GMP Products are intended for research or further manufacturing use only THIS PRODUCT IS NOT INTENDED FOR DIRECT THERAPEUTIC USE IN HUMANS