Learn More
Lonza Walkersville PowerCHO 2 SF Medium 20L bag
Catalog No. 5000496407
Encompass
PowerCHO 2 Serum-free Medium 20 L Bags are hydrolysate-free serum-free and non-animal origin media for supporting high-density CHO cells in suspension For therapeutic bioprocessing applications these protein-free formulations also facilitate both downstream purification and regulatory compliance PowerCHO 2 Serum-free Medium 20 L Bag contains HEPES and Pluronic F-68 but does not contain L-glutamine phenol red hypoxanthine and thymidine PowerCHO 2 Serum-free Medium 20 L bag is a chemically defined serum-free CHO media optimized for both cell growth and protein production The 20 L bag provides a flexible single use approach This BioWhittaker Cell Culture Media is produced according to cGMP and labelled For Further Manufacturing (FFM) GMP Products are intended for research or further manufacturing use only THIS PRODUCT IS NOT INTENDED FOR DIRECT THERAPEUTIC USE IN HUMANS
Catalog No. 50-004-96407 Supplier Lonza Walkersville Supplier No. BE12771P20
PowerCHO 2 Serum-free Medium 20 L Bags are hydrolysate-free serum-free and non-animal origin media for supporting high-density CHO cells in suspension For therapeutic bioprocessing applications these protein-free formulations also facilitate both downstream purification and regulatory compliance PowerCHO 2 Serum-free Medium 20 L Bag contains HEPES and Pluronic F-68 but does not contain L-glutamine phenol red hypoxanthine and thymidine PowerCHO 2 Serum-free Medium 20 L bag is a chemically defined serum-free CHO media optimized for both cell growth and protein production The 20 L bag provides a flexible single use approach This BioWhittaker Cell Culture Media is produced according to cGMP and labelled For Further Manufacturing (FFM) GMP Products are intended for research or further manufacturing use only THIS PRODUCT IS NOT INTENDED FOR DIRECT THERAPEUTIC USE IN HUMANS