Learn More
Lonza Walkersville Polyethylene Glycol for 50L
Catalog No. 5000496255
Encompass
PEG is a supplement of PowerCHO 2 Powder 50 L Kit (sold separately or as part of the kit)This BioWhittaker Cell Culture Media is produced according to cGMP and labelled For Further Manufacturing (FFM) GMP Products are intended for research or further manufacturing use only THIS PRODUCT IS NOT INTENDED FOR DIRECT THERAPEUTIC USE IN HUMANS
Catalog No. 50-004-96255 Supplier Lonza Walkersville Supplier No. VPW070F
PEG is a supplement of PowerCHO 2 Powder 50 L Kit (sold separately or as part of the kit)This BioWhittaker Cell Culture Media is produced according to cGMP and labelled For Further Manufacturing (FFM) GMP Products are intended for research or further manufacturing use only THIS PRODUCT IS NOT INTENDED FOR DIRECT THERAPEUTIC USE IN HUMANS