Learn More
Lonza Walkersville PowerCHO 2 SF Medium 50L
Catalog No. 5000496333
Encompass
PowerCHO Media are hydrolysate-free serum-free and non-animal origin media for supporting high-density CHO cells in suspension For therapeutic bioprocessing applications these protein-free formulations also facilitate both downstream purification and regulatory compliance PowerCHO Medium contains HEPES and Pluronic F-68 but does not contain L-glutamine phenol red hypoxanthine and thymidine PowerCHO 2 Chemically Defined Serum-free CHO Media is optimized for both cell growth and protein production The basal powder needs several components to produce the liquid PowerCHO 2 Medium CHO media can be used in fed-batch cell culturing we provide several feeds like eCHO Feed Power Feed A and Xtreme Feed compatible with PowerCHO 2 Media This BioWhittaker Cell Culture Media is produced according to cGMP and labelled For Further Manufacturing (FFM)
Catalog No. 50-004-96333 Supplier Lonza Walkersville Supplier No. VPW088F
PowerCHO Media are hydrolysate-free serum-free and non-animal origin media for supporting high-density CHO cells in suspension For therapeutic bioprocessing applications these protein-free formulations also facilitate both downstream purification and regulatory compliance PowerCHO Medium contains HEPES and Pluronic F-68 but does not contain L-glutamine phenol red hypoxanthine and thymidine PowerCHO 2 Chemically Defined Serum-free CHO Media is optimized for both cell growth and protein production The basal powder needs several components to produce the liquid PowerCHO 2 Medium CHO media can be used in fed-batch cell culturing we provide several feeds like eCHO Feed Power Feed A and Xtreme Feed compatible with PowerCHO 2 Media This BioWhittaker Cell Culture Media is produced according to cGMP and labelled For Further Manufacturing (FFM)