Learn More
Lonza Walkersville PowerCHO 2 Powder Kit 50L
Catalog No. 5000496411
Encompass
PowerCHO 2 Powder Kit 50 L is hydrolysate-free serum-free and non-animal origin medium for supporting high-density CHO cells in suspension For therapeutic bioprocessing applications these protein-free formulations also facilitate both downstream purification and regulatory compliance PowerCHO 2 Powder Kit 50 L contains HEPES and Pluronic F-68 but does not contain L-glutamine phenol red hypoxanthine and thymidine PowerCHO 2 Powder Kit 50 L is a Chemically Defined Serum-free CHO Media is optimized for both cell growth and protein production The powder kit contains several components to produce the liquid medium PowerCHO 2 Powder Kit 50 L is compatible with fed-batch cell culturing with one of our feeds eCHO Feed PowerFeed A CHO Xtreme Feed This BioWhittaker Cell Culture Media is produced according to cGMP and labelled For Further Manufacturing (FFM)
Catalog No. 50-004-96411 Supplier Lonza Walkersville Supplier No. BE15771NF
PowerCHO 2 Powder Kit 50 L is hydrolysate-free serum-free and non-animal origin medium for supporting high-density CHO cells in suspension For therapeutic bioprocessing applications these protein-free formulations also facilitate both downstream purification and regulatory compliance PowerCHO 2 Powder Kit 50 L contains HEPES and Pluronic F-68 but does not contain L-glutamine phenol red hypoxanthine and thymidine PowerCHO 2 Powder Kit 50 L is a Chemically Defined Serum-free CHO Media is optimized for both cell growth and protein production The powder kit contains several components to produce the liquid medium PowerCHO 2 Powder Kit 50 L is compatible with fed-batch cell culturing with one of our feeds eCHO Feed PowerFeed A CHO Xtreme Feed This BioWhittaker Cell Culture Media is produced according to cGMP and labelled For Further Manufacturing (FFM)