Learn More
Lonza Walkersville ProCHO 5 CHO Medium NAO 50L
Catalog No. 5000496348
Encompass
ProCHO-5 powder Kits (50L) Protein-free CHO media were developed specifically to facilitate the production and downstream processing of recombinant proteins expressed in CHO cells These protein-free formulations support high-density cultures without the need for animal derived components Very low levels of human recombinant insulin facilitate both downstream purification and regulatory compliance ProCHO-5 - For CHO cells already growing in suspension Supports increased protein production Contains 0 1% Pluronic F-68 Does not contain L-glutamine phenol red hypoxanthine or thymidine ProCHO 5 Powder Kits allow production of liquid medium in any required volume This BioWhittaker Cell Culture Media is produced according to cGMP and labelled For Further Manufacturing (FFM) GMP Products are intended for research or further manufacturing use only THIS PRODUCT IS NOT INTENDED FOR DIRECT THERAPEUTIC USE IN HUMANS
Catalog No. 50-004-96348 Supplier Lonza Walkersville Supplier No. BE15766F
ProCHO-5 powder Kits (50L) Protein-free CHO media were developed specifically to facilitate the production and downstream processing of recombinant proteins expressed in CHO cells These protein-free formulations support high-density cultures without the need for animal derived components Very low levels of human recombinant insulin facilitate both downstream purification and regulatory compliance ProCHO-5 - For CHO cells already growing in suspension Supports increased protein production Contains 0 1% Pluronic F-68 Does not contain L-glutamine phenol red hypoxanthine or thymidine ProCHO 5 Powder Kits allow production of liquid medium in any required volume This BioWhittaker Cell Culture Media is produced according to cGMP and labelled For Further Manufacturing (FFM) GMP Products are intended for research or further manufacturing use only THIS PRODUCT IS NOT INTENDED FOR DIRECT THERAPEUTIC USE IN HUMANS