Learn More
Lonza Walkersville PowerCHO 2 SF Medium 500L
Catalog No. 5000496330
Encompass
PowerCHO Media are hydrolysate-free serum-free and non-animal origin media for supporting high-density CHO cells in suspension For therapeutic bioprocessing applications these protein-free formulations also facilitate both downstream purification and regulatory compliance PowerCHO contains HEPES and Pluronic F-68 but does not contain L-glutamine phenol red hypoxanthine and thymidine PowerCHO 2 Chemically Defined Serum-free CHO Media is optimized for both cell growth and protein production The basal powder needs several components to produce the liquid PowerCHO 2 medium CHO media can be used in fed-batch cell culturing we provide several feeds like eCHO Feed Power Feed A and Xtreme Feed compatible with PowerCHO 2 This BioWhittaker Cell Culture Media is produced according to cGMP and labelled For Further Manufacturing (FFM) GMP Products are intended for research or further manufacturing use only
Catalog No. 50-004-96330 Supplier Lonza Walkersville Supplier No. VPW088L
PowerCHO Media are hydrolysate-free serum-free and non-animal origin media for supporting high-density CHO cells in suspension For therapeutic bioprocessing applications these protein-free formulations also facilitate both downstream purification and regulatory compliance PowerCHO contains HEPES and Pluronic F-68 but does not contain L-glutamine phenol red hypoxanthine and thymidine PowerCHO 2 Chemically Defined Serum-free CHO Media is optimized for both cell growth and protein production The basal powder needs several components to produce the liquid PowerCHO 2 medium CHO media can be used in fed-batch cell culturing we provide several feeds like eCHO Feed Power Feed A and Xtreme Feed compatible with PowerCHO 2 This BioWhittaker Cell Culture Media is produced according to cGMP and labelled For Further Manufacturing (FFM) GMP Products are intended for research or further manufacturing use only