Learn More
Lonza Walkersville Ferric Citrate Agent 500mL
Catalog No. 5000496251
Encompass
Anti-clumping Agent A is a buffered concentrated solution of Dextran sulfate and recommended usage is at 100 dilution Anti-clumping Agent B is a buffered concentrated solution of ferric citrate and recommend use it at 100 or 200 dilution As a result of using Anti-clumping Agents A and B from Lonza clumping of cells cultured in serum-free medium is minimized leading to asignificant increase in cell density and productivity This BioWhittaker Cell Culture Media is produced according to cGMP and labelled For Further Manufacturing (FFM) GMP Products are intended for research or further manufacturing use only THIS PRODUCT IS NOT INTENDED FOR DIRECT THERAPEUTIC USE IN HUMANS
Catalog No. 50-004-96251 Supplier Lonza Walkersville Supplier No. BE02059F
Anti-clumping Agent A is a buffered concentrated solution of Dextran sulfate and recommended usage is at 100 dilution Anti-clumping Agent B is a buffered concentrated solution of ferric citrate and recommend use it at 100 or 200 dilution As a result of using Anti-clumping Agents A and B from Lonza clumping of cells cultured in serum-free medium is minimized leading to asignificant increase in cell density and productivity This BioWhittaker Cell Culture Media is produced according to cGMP and labelled For Further Manufacturing (FFM) GMP Products are intended for research or further manufacturing use only THIS PRODUCT IS NOT INTENDED FOR DIRECT THERAPEUTIC USE IN HUMANS