Learn More
Research Products International Corp Tissue Culture Plates, Sterile, 96 Well Array, 0.39ml Well Volume, 100 per Package
SDP
Catalog No. 50998541
Encompass
Tissue Culture Plates are designed for a wide range of cell and tissue culture applications. Manufactured from crystal clear virgin polystyrene and high quality polyethylene to ensure cell integrity. Plates feature raised well rims, specially treated well surface, and alphanumeric indexing for easy identification. Well surface is smooth and free from striation, maximizing growth area, allowing superior cell attachment and growth. Supplied 100 per package, sterile, individually wrapped. Supplied with lid.
Catalog No. 50-998-541 Supplier Research Products International Corp Supplier No. 141368
Tissue Culture Plates are designed for a wide range of cell and tissue culture applications. Manufactured from crystal clear virgin polystyrene and high quality polyethylene to ensure cell integrity. Plates feature raised well rims, specially treated well surface, and alphanumeric indexing for easy identification. Well surface is smooth and free from striation, maximizing growth area, allowing superior cell attachment and growth. Supplied 100 per package, sterile, individually wrapped. Supplied with lid.