Learn More
USA Scientific Inc Culture Tube 14mL, 18x95mm
Catalog No. 502745345
Encompass
Tube 14.0 mL, PP, 18/95 mm, round bottom, two-position vent stopper, natural, graduation, writing area, str, 25/bag, 1000/case
Catalog No. 50-274-5345 Supplier USA Scientific Inc Supplier No. 56187262
Tube 14.0 mL, PP, 18/95 mm, round bottom, two-position vent stopper, natural, graduation, writing area, str, 25/bag, 1000/case