Learn More
USA Scientific Inc Culture Tube, 4.5 mL, 75 mm
Catalog No. 502744940
Encompass
Culture Tube, 4.5 mL, 12.4x 75 mm, str, w Snap (Vent) Cap, PS, round bottom, 80 bags of 25, total 2000 pcs
Catalog No. 50-274-4940 Supplier USA Scientific Inc Supplier No. 56120180
Culture Tube, 4.5 mL, 12.4x 75 mm, str, w Snap (Vent) Cap, PS, round bottom, 80 bags of 25, total 2000 pcs