Learn More
CELLTREAT Scientific Products 5mL Culture Tube and Dual Cap, PP, Sterile
SDP
Catalog No. NC2131750
Encompass_Preferred
Dual caps allow for either vented or sealed positions. Polypropylene tubes offer better chemical resistance. Suitable for FACS and flow cytometry applications. Used for bacterial & suspension culture.
This item is not returnable. View return policy
Catalog No. NC2131750 Supplier CELLTREAT Scientific Products Supplier No. 230433
Dual caps allow for either vented or sealed positions. Polypropylene tubes offer better chemical resistance. Suitable for FACS and flow cytometry applications. Used for bacterial & suspension culture.